Product Details

SNP ID
rs77060031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81312820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTCCCTGCTGGGCGGCAGACC[A/C]CATAGCTGTGCGGGTTCTTCTTCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00482 PubMed Links

Gene Details

Gene
LINC00482
Gene Name
long intergenic non-protein coding RNA 482
There are no transcripts associated with this gene.

Gene
TMEM105
Gene Name
transmembrane protein 105
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178520.3 1771 UTR 3 NP_848615.1
XM_017024522.1 1771 Intron XP_016880011.1

View Full Product Details