Product Details

SNP ID
rs74441216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28901055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTGGCGAGCTCCCGGGCAAAC[A/G]GTACATTAGCCAGCTTAGTGTCAGC
Phenotype
MIM: 616157 MIM: 131560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144683.3 744 Missense Mutation CCG,CTG P206L NP_653284.2
Gene
FLOT2
Gene Name
flotillin 2
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
There are no transcripts associated with this gene.

View Full Product Details