Product Details

SNP ID
rs77926367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32021754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTAGCTGAACGTTGGAGCCTTC[C/T]TGAGATTGTTGGGATTCCACACCAA
Phenotype
MIM: 616558
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC37B PubMed Links

Gene Details

Gene
LRRC37B
Gene Name
leucine rich repeat containing 37B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321350.1 813 Missense Mutation CCT,CTT P121L NP_001308279.1
NM_052888.2 813 Intron NP_443120.2

View Full Product Details