Product Details

SNP ID
rs77778824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12311873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACAGGATAGGTGGCAGATTTGG[A/C]AGTTTCCCACCTATTTTAATGTTAG
Phenotype
MIM: 615103
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TUBB6 PubMed Links

Gene Details

Gene
TUBB6
Gene Name
tubulin beta 6 class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303524.1 Intron NP_001290453.1
NM_001303525.1 Intron NP_001290454.1
NM_001303526.1 Intron NP_001290455.1
NM_001303527.1 Intron NP_001290456.1
NM_001303528.1 Intron NP_001290457.1
NM_001303529.1 Intron NP_001290458.1
NM_001303530.1 Intron NP_001290459.1
NM_032525.2 Intron NP_115914.1

View Full Product Details