Product Details
- SNP ID
-
rs75423671
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:3266437 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATATCTTATTGTCACAATATCTTA[C/T]TGGTTACACGGGTCAACACTTTTCA
- Phenotype
-
MIM: 609211
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC104968399
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140337632] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC104968399
- Gene Name
- uncharacterized LOC104968399
There are no transcripts associated with this gene.
- Gene
- MYL12B
- Gene Name
- myosin light chain 12B
View Full Product Details