Product Details

SNP ID
rs78801230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:51036482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTTAATGGGGTAAGCTAAGCC[A/T]GGAATCATTGTACATAAACACATTA
Phenotype
MIM: 600993
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SMAD4 PubMed Links

Gene Details

Gene
SMAD4
Gene Name
SMAD family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005359.5 Intron NP_005350.1

View Full Product Details