Product Details

SNP ID
rs76681915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21651167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTATAAATACCCTTAGGAATGT[A/G]CCACCTTATAACTTACATTGAAATA
Phenotype
MIM: 612197
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD3 PubMed Links

Gene Details

Gene
ABHD3
Gene Name
abhydrolase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308256.1 1794 UTR 3 NP_001295185.1
NM_001308257.1 1794 UTR 3 NP_001295186.1
NM_138340.4 1794 UTR 3 NP_612213.2
XM_011525847.2 1794 Intron XP_011524149.1
XM_017025571.1 1794 UTR 3 XP_016881060.1
XM_017025572.1 1794 UTR 3 XP_016881061.1
XM_017025573.1 1794 UTR 3 XP_016881062.1
XM_017025574.1 1794 UTR 3 XP_016881063.1

View Full Product Details