Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040283.2 | Intron | NP_001035373.1 | |||
XM_011527214.1 | Intron | XP_011525516.1 | |||
XM_011527215.1 | Intron | XP_011525517.1 | |||
XM_011527216.2 | Intron | XP_011525518.1 | |||
XM_017027123.1 | Intron | XP_016882612.1 | |||
XM_017027124.1 | Intron | XP_016882613.1 | |||
XM_017027125.1 | Intron | XP_016882614.1 | |||
XM_017027126.1 | Intron | XP_016882615.1 | |||
XM_017027127.1 | Intron | XP_016882616.1 | |||
XM_017027128.1 | Intron | XP_016882617.1 | |||
XM_017027129.1 | Intron | XP_016882618.1 |