Product Details

SNP ID
rs79673670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48882222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTCCTAGAAGGTAAAGAAGGG[A/G]CTGTGGTTTTCTCAGAGGCTTTCGA
Phenotype
MIM: 611048 MIM: 602309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R15A PubMed Links

Gene Details

Gene
PPP1R15A
Gene Name
protein phosphatase 1 regulatory subunit 15A
There are no transcripts associated with this gene.

Gene
TULP2
Gene Name
tubby like protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003323.2 Intron NP_003314.2
XM_006723347.3 Intron XP_006723410.1
XM_006723348.3 Intron XP_006723411.1
XM_011527252.2 Intron XP_011525554.1
XM_011527253.2 Intron XP_011525555.1
XM_011527254.2 Intron XP_011525556.1
XM_011527255.2 Intron XP_011525557.1
XM_011527256.2 Intron XP_011525558.1

View Full Product Details