Product Details

SNP ID
rs76397153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49128441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAGGGAGGAAGAGATTGCAGA[A/G]CTGAAGGCGGAACGGAACAACACGC
Phenotype
MIM: 612331 MIM: 603144
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf73 PubMed Links

Gene Details

Gene
C19orf73
Gene Name
chromosome 19 open reading frame 73
There are no transcripts associated with this gene.

Gene
LIN7B
Gene Name
lin-7 homolog B, crumbs cell polarity complex component
There are no transcripts associated with this gene.

Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 664 Silent Mutation GAA,GAG E105E NP_003651.1
XM_017027407.1 664 Silent Mutation GAA,GAG E105E XP_016882896.1

View Full Product Details