Product Details

SNP ID
rs74491170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49581063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCCCAGGGACTAAGGCATATC[A/G]TTCCCGCTAGTTTCTAGATACCTAG
Phenotype
MIM: 616759 MIM: 604429
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOSIP PubMed Links

Gene Details

Gene
NOSIP
Gene Name
nitric oxide synthase interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270960.1 Intron NP_001257889.1
NM_015953.4 Intron NP_057037.1
XM_005258964.4 Intron XP_005259021.1
XM_006723235.3 Intron XP_006723298.1
XM_011527015.2 Intron XP_011525317.1
XM_011527017.2 Intron XP_011525319.1
XM_017026851.1 Intron XP_016882340.1
XM_017026852.1 Intron XP_016882341.1
Gene
PRRG2
Gene Name
proline rich and Gla domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000951.2 Intron NP_000942.1
NM_001316335.1 Intron NP_001303264.1
XM_006723286.2 Intron XP_006723349.1
XM_011527124.1 Intron XP_011525426.1

View Full Product Details