Product Details

SNP ID
rs77714959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49773611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTCCTGCCCTAGGGTGCTCACG[C/T]TCTAGAGAGGAGATGGTCCACTACA
Phenotype
MIM: 601026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP2A1 PubMed Links

Gene Details

Gene
AP2A1
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014203.2 Intron NP_055018.2
NM_130787.2 Intron NP_570603.2
XM_011526556.2 Intron XP_011524858.1
XM_011526557.2 Intron XP_011524859.1

View Full Product Details