Product Details

SNP ID
rs78569821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51493705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTATTGGCAATTCTCATAAGTCT[A/C]ACTTCAGAATTTACCCATAAGACAG
Phenotype
MIM: 606094
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
There are no transcripts associated with this gene.

Gene
SIGLEC12
Gene Name
sialic acid binding Ig like lectin 12 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033329.2 Intron NP_201586.1
NM_053003.3 Intron NP_443729.1
XM_011527441.2 Intron XP_011525743.1
XM_011527442.2 Intron XP_011525744.1

View Full Product Details