Product Details

SNP ID
rs75422986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52193041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCATCTGCGTAACAAATTATTGG[A/G]TGCTTGCTCTATGTTAGGTACAGTA
Phenotype
MIM: 605983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP2R1A PubMed Links

Gene Details

Gene
PPP2R1A
Gene Name
protein phosphatase 2 scaffold subunit Aalpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014225.5 Intron NP_055040.2
XM_017026929.1 Intron XP_016882418.1

View Full Product Details