Product Details
- SNP ID
-
rs77493998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55442223 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGTTGATATCCCGGTGCGCCCGG[G/T]GCGCACGGGACGCCTTGCTGAAGGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SHISA7
PubMed Links
Gene Details
- Gene
- SHISA7
- Gene Name
- shisa family member 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145176.1 |
641 |
Missense Mutation |
CAC,CCC |
H214P |
NP_001138648.1 |
View Full Product Details