Product Details

SNP ID
rs74258046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56180271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGTGGGCACGATGTTGTGCCA[C/T]AGAGCTCTAGGACTTCTTCATCCTG
Phenotype
MIM: 611178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALP PubMed Links

Gene Details

Gene
GALP
Gene Name
galanin like peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145546.1 Intron NP_001139018.1
NM_033106.3 Intron NP_149097.1
Gene
ZSCAN5B
Gene Name
zinc finger and SCAN domain containing 5B
There are no transcripts associated with this gene.

View Full Product Details