Product Details
- SNP ID
-
rs76795464
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57530185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCAGTCACCAGAAAAATCAAGTGA[C/T]TATAGAATTTGAACTTTGAGCCCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF549
PubMed Links
Gene Details
- Gene
- ZNF549
- Gene Name
- zinc finger protein 549
View Full Product Details