Product Details

SNP ID
rs75763025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAATCTCCCCAGTGGGAATGTCC[A/G]TATCTATGAAGTTTCACTTCTTACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2471 UTR 3 NP_001308304.1
NM_001321376.1 2471 UTR 3 NP_001308305.1
NM_024833.2 2471 UTR 3 NP_079109.2
XM_017027314.1 2471 UTR 3 XP_016882803.1

View Full Product Details