Product Details

SNP ID
rs79204341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20004639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGTACTTTCTTCACACACCTC[A/G]TATTTCAGTGTCCTCAATCTTCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF682 PubMed Links

Gene Details

Gene
ZNF682
Gene Name
zinc finger protein 682
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077349.1 3090 Intron NP_001070817.1
NM_033196.2 3090 Intron NP_149973.1
XM_017027455.1 3090 UTR 3 XP_016882944.1
XM_017027456.1 3090 Intron XP_016882945.1

View Full Product Details