Product Details

SNP ID
rs79368249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12904473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCATAAGGAATCCCCCCTCTC[A/G]CTGGGTGGAGTCTGTCACCTTGGTC
Phenotype
MIM: 608801 MIM: 611487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCDH PubMed Links

Gene Details

Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000159.3 Intron NP_000150.1
NM_013976.3 Intron NP_039663.1
XM_006722721.2 Intron XP_006722784.1
XM_011527899.2 Intron XP_011526201.1
XM_011527900.2 Intron XP_011526202.1
XM_017026580.1 Intron XP_016882069.1
Gene
SYCE2
Gene Name
synaptonemal complex central element protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105578.1 Intron NP_001099048.1
XM_005259848.4 Intron XP_005259905.1
XM_011527882.2 Intron XP_011526184.1
XM_011527883.2 Intron XP_011526185.1

View Full Product Details