Product Details

SNP ID
rs79484951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15794090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTCCGTGTCTGAATTCATCCT[C/T]GTTGGCTTCTCTGCCTTCCCCCACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10H5 PubMed Links

Gene Details

Gene
OR10H5
Gene Name
olfactory receptor family 10 subfamily H member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004466.1 42 Silent Mutation CTC,CTT L14L NP_001004466.1

View Full Product Details