Product Details

SNP ID
rs80068367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32443679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAAAACATGTCTAAATTATGC[C/T]AGAGGACTAAAGACCTCTTGGGTTG
Phenotype
MIM: 612990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASXL1 PubMed Links

Gene Details

Gene
ASXL1
Gene Name
additional sex combs like 1, transcriptional regulator
There are no transcripts associated with this gene.

Gene
NOL4L
Gene Name
nucleolar protein 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256798.1 5482 UTR 3 NP_001243727.1
NM_080616.4 5482 UTR 3 NP_542183.2
XM_005260282.4 5482 Intron XP_005260339.1
XM_005260283.4 5482 Intron XP_005260340.1
XM_005260284.4 5482 Intron XP_005260341.1
XM_005260285.4 5482 Intron XP_005260342.2
XM_005260286.4 5482 Intron XP_005260343.1
XM_005260288.2 5482 Intron XP_005260345.1
XM_005260289.4 5482 Intron XP_005260346.1
XM_006723702.3 5482 Intron XP_006723765.1
XM_006723703.3 5482 Intron XP_006723766.1
XM_011528562.2 5482 Intron XP_011526864.1
XM_011528563.1 5482 Intron XP_011526865.1
XM_017027669.1 5482 Intron XP_016883158.1
XM_017027670.1 5482 Intron XP_016883159.1

View Full Product Details