Product Details

SNP ID
rs79781066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44480301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGGGCCCTGCAGCTCCTCGTCA[A/G]CTACCACAGCTGTAGAAGAAGCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTPAL PubMed Links

Gene Details

Gene
TTPAL
Gene Name
alpha tocopherol transfer protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039199.2 438 Missense Mutation AAC,AGC N101S NP_001034288.1
NM_001261839.1 438 Missense Mutation AAC,AGC N101S NP_001248768.1
NM_024331.4 438 Missense Mutation AAC,AGC N101S NP_077307.2
XM_011529043.2 438 Missense Mutation AAC,AGC N101S XP_011527345.1
XM_011529044.2 438 Missense Mutation AAC,AGC N101S XP_011527346.1
XM_011529045.2 438 Missense Mutation AAC,AGC N101S XP_011527347.1
XM_017028063.1 438 Missense Mutation AAC,AGC N101S XP_016883552.1

View Full Product Details