Product Details

SNP ID
rs74715401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44943615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCATTTCAGAGGTTTCAGTCAC[A/G]TCTTATGGGCCACCCACATGCCACG
Phenotype
MIM: 616049
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PABPC1L PubMed Links

Gene Details

Gene
PABPC1L
Gene Name
poly(A) binding protein cytoplasmic 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001124756.2 Intron NP_001118228.1
Gene
TOMM34
Gene Name
translocase of outer mitochondrial membrane 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006809.4 Intron NP_006800.2
XM_005260254.2 Intron XP_005260311.1
XM_011528501.1 Intron XP_011526803.1
XM_017027600.1 Intron XP_016883089.1

View Full Product Details