Product Details

SNP ID
rs79493196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45833926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCGGCGCGCCCCTGAATGCAC[C/G]GTGGGACGCAGGAGGTAGAGGCGCA
Phenotype
MIM: 608123 MIM: 191039
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACOT8 PubMed Links

Gene Details

Gene
ACOT8
Gene Name
acyl-CoA thioesterase 8
There are no transcripts associated with this gene.

Gene
SNX21
Gene Name
sorting nexin family member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042632.2 96 Missense Mutation CGT,GGT R3G NP_001036097.1
NM_001042633.2 96 Missense Mutation CGT,GGT R3G NP_001036098.1
NM_033421.3 96 Missense Mutation CGT,GGT R3G NP_219489.1
NM_152897.2 96 Missense Mutation CGT,GGT R3G NP_690857.1
XM_005260608.4 96 Intron XP_005260665.1
XM_011529095.2 96 Intron XP_011527397.1
XM_011529096.2 96 Missense Mutation CGT,GGT R3G XP_011527398.1
XM_011529098.2 96 Missense Mutation CGT,GGT R3G XP_011527400.1
XM_011529099.2 96 Missense Mutation CGT,GGT R3G XP_011527401.1
XM_017028120.1 96 Missense Mutation CGT,GGT R3G XP_016883609.1
Gene
TNNC2
Gene Name
troponin C2, fast skeletal type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003279.2 96 Intron NP_003270.1
XM_011529031.2 96 Intron XP_011527333.1

View Full Product Details