Product Details

SNP ID
rs77850827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62309991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCACAGGAGGGGCCTCACCCG[A/C]TAGCCGGTGCCACTGGCCATCACAC
Phenotype
MIM: 610650 MIM: 601033
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADRM1 PubMed Links

Gene Details

Gene
ADRM1
Gene Name
adhesion regulating molecule 1
There are no transcripts associated with this gene.

Gene
LAMA5
Gene Name
laminin subunit alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005560.4 10772 Missense Mutation NP_005551.3
XM_006723796.3 10772 Missense Mutation XP_006723859.1
XM_006723798.3 10772 Intron XP_006723861.1
XM_011528818.2 10772 Missense Mutation XP_011527120.1
XM_011528819.2 10772 Missense Mutation XP_011527121.1

View Full Product Details