Product Details

SNP ID
rs79979540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36329565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGGCCTCCACCAAGCTTTTGGT[C/T]CTGACACTTCCTCTCTATCCCTGAT
Phenotype
MIM: 616191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLGAP4 PubMed Links

Gene Details

Gene
DLGAP4
Gene Name
DLG associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042486.3 Intron NP_001035951.1
NM_014902.5 Intron NP_055717.2
NM_183006.3 Intron NP_892118.1
XM_005260332.4 Intron XP_005260389.1
XM_005260333.4 Intron XP_005260390.1
XM_011528688.2 Intron XP_011526990.1
XM_011528689.2 Intron XP_011526991.1
XM_011528691.2 Intron XP_011526993.1
XM_011528692.2 Intron XP_011526994.1
XM_017027725.1 Intron XP_016883214.1
XM_017027726.1 Intron XP_016883215.1
XM_017027727.1 Intron XP_016883216.1

View Full Product Details