Product Details

SNP ID
rs74932642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:16748445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCTCCCGGGTCTTGTGGCTG[T/G]ATGTGCTGTGCATGGAATATTTATG
Phenotype
MIM: 606067 MIM: 603520
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
OTOR PubMed Links

Gene Details

Gene
OTOR
Gene Name
otoraplin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020157.3 93 Missense Mutation GGA,GTA G15V NP_064542.1
XM_017027959.1 93 Missense Mutation GGA,GTA G15V XP_016883448.1
Gene
SNRPB2
Gene Name
small nuclear ribonucleoprotein polypeptide B2
There are no transcripts associated with this gene.

View Full Product Details