Product Details

SNP ID
rs75221813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3855633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGTCCTCAGATTTCTCAGGA[A/G]CCCCATTGTTGTCTCCGCTGTCCTC
Phenotype
MIM: 609676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAVS PubMed Links

Gene Details

Gene
MAVS
Gene Name
mitochondrial antiviral signaling protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206491.1 Intron NP_001193420.1
NM_020746.4 Intron NP_065797.2

View Full Product Details