Product Details

SNP ID
rs79552090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:7884536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGTACAATTTATGTGAACACA[C/T]GAAAGATGAGAAGTTTTTCATGTAG
Phenotype
MIM: 605023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAO1 PubMed Links

Gene Details

Gene
HAO1
Gene Name
hydroxyacid oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017545.2 Intron NP_060015.1

View Full Product Details