Product Details

SNP ID
rs79296943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33652197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGGACCACGAGCACTGAAGGTG[A/G]CAGGACGGTCCAGGGCCAGTCAACC
Phenotype
MIM: 602442
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITSN1 PubMed Links

Gene Details

Gene
ITSN1
Gene Name
intersectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001132.1 Intron NP_001001132.1
NM_003024.2 Intron NP_003015.2
XM_005261026.2 Intron XP_005261083.1
XM_005261027.1 Intron XP_005261084.1
XM_005261031.1 Intron XP_005261088.1
XM_011529691.1 Intron XP_011527993.1
XM_011529692.1 Intron XP_011527994.1
XM_011529693.2 Intron XP_011527995.1
XM_017028428.1 Intron XP_016883917.1
XM_017028429.1 Intron XP_016883918.1
XM_017028430.1 Intron XP_016883919.1
XM_017028431.1 Intron XP_016883920.1
XM_017028432.1 Intron XP_016883921.1
XM_017028433.1 Intron XP_016883922.1
XM_017028434.1 Intron XP_016883923.1
XM_017028435.1 Intron XP_016883924.1
XM_017028436.1 Intron XP_016883925.1
XM_017028437.1 Intron XP_016883926.1
XM_017028438.1 Intron XP_016883927.1
XM_017028439.1 Intron XP_016883928.1
XM_017028440.1 Intron XP_016883929.1
XM_017028441.1 Intron XP_016883930.1

View Full Product Details