Product Details

SNP ID
rs79135244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37225182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCATTAGCAAGTCAGAGGTTTC[C/T]TTTTGTCCCCATGATGCCACTCTCC
Phenotype
MIM: 605298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCR3 PubMed Links

Gene Details

Gene
DSCR3
Gene Name
DSCR3 arrestin fold containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006052.1 2005 UTR 3 NP_006043.1
XM_005260909.4 2005 UTR 3 XP_005260966.1
XM_005260911.4 2005 UTR 3 XP_005260968.1
XM_011529423.2 2005 UTR 3 XP_011527725.1
Gene
DSCR9
Gene Name
Down syndrome critical region 9 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details