Product Details

SNP ID
rs75506502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45004610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCGGGCAGGCCTCGCCTCTG[C/T]GCGTTTCCCATGGATGAGGTCATAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PICSAR PubMed Links

Gene Details

Gene
PICSAR
Gene Name
P38 inhibited cutaneous squamous cell carcinoma associated lincRNA
There are no transcripts associated with this gene.

View Full Product Details