Product Details

SNP ID
rs74276417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17179796 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGTGTACCTCTAAACGCACTG[C/T]GTTGTAAGCAAAGGAGTGACCTGAT
Phenotype
MIM: 607575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CECR1 PubMed Links

Gene Details

Gene
CECR1
Gene Name
cat eye syndrome chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282225.1 3374 UTR 3 NP_001269154.1
NM_001282226.1 3374 UTR 3 NP_001269155.1
NM_001282227.1 3374 UTR 3 NP_001269156.1
NM_001282228.1 3374 UTR 3 NP_001269157.1
NM_001282229.1 3374 UTR 3 NP_001269158.1
NM_177405.2 3374 UTR 3 NP_803124.1
XM_006724080.3 3374 UTR 3 XP_006724143.1
XM_011546133.1 3374 UTR 3 XP_011544435.1

View Full Product Details