Product Details

SNP ID
rs75091182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32388037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCTCAGTTTAATGGCTTTCTT[G/T]CTGATTCCAGCATCATGGCAGGTAT
Phenotype
MIM: 613901
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC339666 PubMed Links

Gene Details

Gene
LOC339666
Gene Name
uncharacterized LOC339666
There are no transcripts associated with this gene.

Gene
RTCB
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014306.4 1604 Missense Mutation AGA,AGC R491S NP_055121.1

View Full Product Details