Product Details

SNP ID
rs77905643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21643933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAAGACGGCGAGGGCGACGAC[C/G]TGGACCTATGGACAGTGCGCTGCTC
Phenotype
MIM: 613682 MIM: 607551
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC116 PubMed Links

Gene Details

Gene
CCDC116
Gene Name
coiled-coil domain containing 116
There are no transcripts associated with this gene.

Gene
MIR130B
Gene Name
microRNA 130b
There are no transcripts associated with this gene.

Gene
MIR301B
Gene Name
microRNA 301b
There are no transcripts associated with this gene.

Gene
SDF2L1
Gene Name
stromal cell derived factor 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022044.2 508 Missense Mutation CTG,GTG L142V NP_071327.2

View Full Product Details