Product Details

SNP ID
rs78880044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36516088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCAATGAAAGAAGAGGTCAAA[A/G]AGAAGAGCGGCCAGACTTACAGGAA
Phenotype
MIM: 603915 MIM: 613777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF3D PubMed Links

Gene Details

Gene
EIF3D
Gene Name
eukaryotic translation initiation factor 3 subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003753.3 Intron NP_003744.1
Gene
FOXRED2
Gene Name
FAD dependent oxidoreductase domain containing 2
There are no transcripts associated with this gene.

View Full Product Details