Product Details

SNP ID
rs74729565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37474575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGGTGGGAGTGAAAGAGGGG[A/G]CTGGGCTGCAGGCGGCCGCCCAGCT
Phenotype
MIM: 602577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFNG PubMed Links

Gene Details

Gene
MFNG
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166343.1 918 Silent Mutation NP_001159815.1
NM_002405.3 918 Silent Mutation NP_002396.2

View Full Product Details