Product Details

SNP ID
rs78290806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43418924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCTTTCAGGCAGGAGAGGAGG[G/T]GTTGCTTGAAAATCCCTTCTGCATT
Phenotype
MIM: 602112
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MPPED1 PubMed Links

Gene Details

Gene
MPPED1
Gene Name
metallophosphoesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044370.1 Intron NP_001037835.1
XM_011530367.2 Intron XP_011528669.1

View Full Product Details