Product Details

SNP ID
rs76625778
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26600982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCAAGTAAGTTCCTTGAAGGTA[A/G]GAACTTGGCTGTATTCATCGCTGCT
Phenotype
MIM: 123631 MIM: 600929
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CRYBA4 PubMed Links

Gene Details

Gene
CRYBA4
Gene Name
crystallin beta A4
There are no transcripts associated with this gene.

Gene
CRYBB1
Gene Name
crystallin beta B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001887.3 Intron NP_001878.1
XM_011529899.2 Intron XP_011528201.1

View Full Product Details