Product Details

SNP ID
rs79635048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72184558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCCCAAAGCTGATGCCAGAGT[C/T]GTCAGGAAATCAAAGGCAGTGAGGT
Phenotype
MIM: 300252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIN4 PubMed Links

Gene Details

Gene
PIN4
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170747.1 Intron NP_001164218.1
NM_006223.3 Intron NP_006214.2

View Full Product Details