Product Details
- SNP ID
-
rs75965644
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:183648336 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTAACCTGACATAGTAGCACAC[A/G]CATAGATACATACATAAGTGTAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD4
PubMed Links
Gene Details
- Gene
- RWDD4
- Gene Name
- RWD domain containing 4
View Full Product Details