Product Details

SNP ID
rs77582481
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86883353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTCAGGTCAGATAGTTTGGCA[A/G]GTAGTACATATATATTGTAAGAATA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C4orf36 PubMed Links

Gene Details

Gene
C4orf36
Gene Name
chromosome 4 open reading frame 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144645.3 Intron NP_653246.1
XM_011531610.2 Intron XP_011529912.1
XM_011531611.2 Intron XP_011529913.1
XM_011531612.2 Intron XP_011529914.1
XM_011531613.2 Intron XP_011529915.1
XM_011531615.2 Intron XP_011529917.1
XM_017007746.1 Intron XP_016863235.1

View Full Product Details