Product Details

SNP ID
rs79633135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:87822273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGAAGCCCAATCACCTGACTC[A/G]TCATGCTTTCCAGGATAACCCATCT
Phenotype
MIM: 147563 MIM: 605912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IBSP PubMed Links

Gene Details

Gene
IBSP
Gene Name
integrin binding sialoprotein
There are no transcripts associated with this gene.

Gene
MEPE
Gene Name
matrix extracellular phosphoglycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184694.2 Intron NP_001171623.1
NM_001184695.1 Intron NP_001171624.1
NM_001184696.1 Intron NP_001171625.1
NM_001184697.1 Intron NP_001171626.1
NM_001291183.1 Intron NP_001278112.1
NM_020203.3 Intron NP_064588.1
XM_006714278.2 Intron XP_006714341.1

View Full Product Details