Product Details

SNP ID
rs79504180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTACCTGTGGCTGCACCTCCGGG[A/G]GACCAGGATCACCCGGCTCCCGGAG
Phenotype
MIM: 138297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 755 Missense Mutation GAG,GGG E138G NP_001968.3
XM_017007877.1 755 Intron XP_016863366.1

View Full Product Details