Product Details

SNP ID
rs78047690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:120060921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTCACCGTAGCTGTGATCTGT[C/T]TGATCACTGAACGGATTTCATCCTG
Phenotype
MIM: 601467
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAD2L1 PubMed Links

Gene Details

Gene
MAD2L1
Gene Name
MAD2 mitotic arrest deficient-like 1 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002358.3 Intron NP_002349.1

View Full Product Details