Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001013415.1 | 4113 | UTR 3 | NP_001013433.1 | ||
NM_001257069.1 | 4113 | Intron | NP_001243998.1 | ||
NM_018315.4 | 4113 | UTR 3 | NP_060785.2 | ||
NM_033632.3 | 4113 | UTR 3 | NP_361014.1 | ||
XM_011532083.1 | 4113 | UTR 3 | XP_011530385.1 | ||
XM_011532084.1 | 4113 | UTR 3 | XP_011530386.1 | ||
XM_011532085.1 | 4113 | UTR 3 | XP_011530387.1 | ||
XM_011532086.1 | 4113 | UTR 3 | XP_011530388.1 | ||
XM_011532087.1 | 4113 | UTR 3 | XP_011530389.1 | ||
XM_011532088.1 | 4113 | UTR 3 | XP_011530390.1 | ||
XM_017008362.1 | 4113 | UTR 3 | XP_016863851.1 |