Product Details

SNP ID
rs79175912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136011699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCGTCCACCTCCTCGCCGGC[A/G]TCGAAGGCGGGGTTGGCGGCGGCAC
Phenotype
MIM: 615786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NACC2 PubMed Links

Gene Details

Gene
NACC2
Gene Name
NACC family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144653.4 1737 Silent Mutation GAC,GAT D527D NP_653254.1
XM_011518221.2 1737 Silent Mutation GAC,GAT D527D XP_011516523.1

View Full Product Details