Product Details

SNP ID
rs78045747
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:9105580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGACCCTGAGGCCGATGAAG[A/C]TGAGCGGGATGAGCACCAGCTTCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR157 PubMed Links

Gene Details

Gene
GPR157
Gene Name
G protein-coupled receptor 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024980.4 751 Missense Mutation AGC,ATC S233I NP_079256.4
XM_005263496.4 751 Missense Mutation AGC,ATC S220I XP_005263553.1
XM_005263497.4 751 Intron XP_005263554.1

View Full Product Details