Product Details

SNP ID
rs75973521
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:11937584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGTGCCTGCATCCCAGACCAT[C/T]CCAGGAGAGAAACCGCAGCTGCCCA
Phenotype
MIM: 153454
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLOD1 PubMed Links

Gene Details

Gene
PLOD1
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000302.3 Intron NP_000293.2
NM_001316320.1 Intron NP_001303249.1

View Full Product Details